MAT062353102 Matrix Scientific

[2-(3,5-Dimethyl-1H-pyrazol-1-yl)ethyl]amine dihydrochloride

  • MDL number MFCD06800633
  • Cas Number : 1185304-68-0
  • Molecular Weight : 212.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/016540.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/016540.html
Availability details for
Quantity
  • Availability
  • Add to Cart