MAT062353105 Matrix Scientific

[(3,5-Dimethyl-1H-pyrazol-4-yl)methyl]methylaminedihydrochloride

  • MDL number MFCD06739052
  • Cas Number : 1240289-23-9
  • Molecular Weight : 212.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/016456.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/016456.html
Availability details for
Quantity
  • Availability
  • Add to Cart