MAT062353108 Matrix Scientific

3-(3,5-Dimethyl-pyrazol-1-yl)-propylaminetartrate

  • MDL number MFCD06800638
  • Cas Number : 898272-44-1
  • Molecular Weight : 303.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/011330.html

Safety Information

MSDS http://www.matrixscientific.com/011330.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart