MAT062353180 Matrix Scientific

1-Ethyl-4-piperidin-4-yl-piperazinehydrochloride

  • MDL number MFCD06800708
  • Cas Number : 202991-77-3
  • Molecular Weight : 233.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/008591.html

Safety Information

MSDS http://www.matrixscientific.com/008591.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart