MAT062353195 Matrix Scientific

(4-Fluoro-benzyl)-(2-methoxy-1-methyl-ethyl)-aminehydrochloride

  • MDL number MFCD06739458
  • Cas Number : 418786-13-7
  • Molecular Weight : 233.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010966.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/010966.html
Availability details for
Quantity
  • Availability
  • Add to Cart