MAT062353210 Matrix Scientific

6-Fluoro-1,7-dimethyl-1H-indole-2-carbaldehyde

  • MDL number MFCD06800737
  • Cas Number : 883531-02-0
  • Molecular Weight : 191.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/016216.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/016216.html
Availability details for
Quantity
  • Availability
  • Add to Cart