MAT062353221 Matrix Scientific

6-Fluoro-7-methyl-1H-indole-2-carbaldehyde

  • MDL number MFCD06800751
  • Cas Number : 883531-54-2
  • Molecular Weight : 177.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/016212.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/016212.html
Availability details for
Quantity
  • Availability
  • Add to Cart