MAT062353239 Matrix Scientific

({1-[2-(3-Fluorophenyl)ethyl]piperidin-4-yl}-methyl)methylamine

  • MDL number MFCD06800767
  • Cas Number : 883542-92-5
  • Molecular Weight : 250.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/016436.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/016436.html
Availability details for
Quantity
  • Availability
  • Add to Cart