MAT062353268 Matrix Scientific

2-Hydrazino-nicotinic acid hydrochloride

  • MDL number MFCD06800796
  • Cas Number : 435342-14-6
  • Molecular Weight : 189.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010371.html

Safety Information

MSDS http://www.matrixscientific.com/010371.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart