MAT062353279 Matrix Scientific

2-Iodo-3-nitronaphthalene

  • MDL number MFCD06656539
  • Cas Number : 102153-71-9
  • Molecular Weight : 299.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.matrixscientific.com/016644.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/016644.html
Availability details for
Quantity
  • Availability
  • Add to Cart