MAT062353284 Matrix Scientific

1-(4-Isobutylbenzyl)piperidin-3-aminesulfate

  • MDL number MFCD06800809
  • Cas Number : 1019331-38-4
  • Molecular Weight : 344.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/016422.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/016422.html
Availability details for
Quantity
  • Availability
  • Add to Cart