MAT062353337 Matrix Scientific

(2-Isopropyl-benzoimidazol-1-yl)-acetic acidhydrochloride

  • MDL number MFCD06800862
  • Cas Number : 1185293-03-1
  • Molecular Weight : 254.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/012107.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/012107.html
Availability details for
Quantity
  • Availability
  • Add to Cart