MAT062353340 Matrix Scientific

(4-Isopropylbenzyl)hydrazinedihydrochloride

  • MDL number MFCD06800865
  • Cas Number : 1185304-29-3
  • Molecular Weight : 237.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/016136.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/016136.html
Availability details for
Quantity
  • Availability
  • Add to Cart