MAT062353366 Matrix Scientific

1-Isopropyl-piperazine hydrochloride

  • MDL number MFCD06800891
  • Cas Number : 137186-14-2
  • Molecular Weight : 164.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/008554.html

Safety Information

MSDS http://www.matrixscientific.com/008554.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart