MAT062353421 Matrix Scientific

1-(2-Chloroethyl)piperidine-4-carbonitrilehydrochloride

  • MDL number MFCD00203877
  • Cas Number : 111041-03-3
  • Molecular Weight : 209.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/016500.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/016500.html
Availability details for
Quantity
  • Availability
  • Add to Cart