MAT062353435 Matrix Scientific

1,2-Diiodo-3,4,5,6-tetramethylbenzene

  • MDL number MFCD06656523
  • Cas Number : 5503-82-2
  • Molecular Weight : 386.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.matrixscientific.com/016577.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/016577.html
Availability details for
Quantity
  • Availability
  • Add to Cart