MAT062353438 Matrix Scientific

Cyclohexylmethyl-(tetrahydro-furan-2-ylmethyl)-amine hydrochloride

  • MDL number MFCD03904906
  • Cas Number : 1048649-17-7
  • Molecular Weight : 233.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010399.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/010399.html
Availability details for
Quantity
  • Availability
  • Add to Cart