MAT062353442 Matrix Scientific

2-Chloro-1H-indole-3-carbaldehyde

  • MDL number MFCD00111620
  • Cas Number : 5059-30-3
  • Molecular Weight : 179.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 223-225
Country of Origin USA
Product Link http://www.matrixscientific.com/016480.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/016480.html
Availability details for
Quantity
  • Availability
  • Add to Cart