MAT062353448 Matrix Scientific

But-2-ynyl-isobutyl-amine hydrochloride

  • MDL number MFCD03632849
  • Cas Number : 1185302-04-8
  • Molecular Weight : 161.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010893.html

Safety Information

MSDS http://www.matrixscientific.com/010893.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart