MAT062353465 Matrix Scientific

1-(2,5-Dimethoxy-benzyl)-piperazinehydrochloride

  • MDL number MFCD03093088
  • Cas Number : 374897-99-1
  • Molecular Weight : 272.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/008147.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/008147.html
Availability details for
Quantity
  • Availability
  • Add to Cart