MAT062353466 Matrix Scientific

[(5-Chloro-1H-indol-2-yl)methyl]-methylamine

  • MDL number MFCD05181972
  • Cas Number : 883528-38-9
  • Molecular Weight : 194.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/016184.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/016184.html
Availability details for
Quantity
  • Availability
  • Add to Cart