MAT062353467 Matrix Scientific

1-(3,4-Dimethoxy-benzyl)-piperazinetrifluoroacetate

  • MDL number MFCD02052086
  • Cas Number : 471916-70-8
  • Molecular Weight : 350.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/008080.html

Safety Information

MSDS http://www.matrixscientific.com/008080.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart