MAT062353499 Matrix Scientific

4-(4-Fluoro-phenylamino)-butan-1-olhydrochloride

  • MDL number MFCD06662286
  • Cas Number : 435345-40-7
  • Molecular Weight : 219.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/008304.html

Safety Information

MSDS http://www.matrixscientific.com/008304.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart