MAT062353504 Matrix Scientific

2-Chloro-N-[2-(5-methyl-furan-2-yl)-phenyl]-acetamide

  • MDL number MFCD02996407
  • Cas Number : 381681-57-8
  • Molecular Weight : 249.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/016684.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/016684.html
Availability details for
Quantity
  • Availability
  • Add to Cart