MAT062353539 Matrix Scientific

1,3-Dimethyl-benzo[4,5]imidazo-[1,2-a]pyridine-4-carboxylic acid

  • MDL number MFCD00839716
  • Cas Number : 10326-57-5
  • Molecular Weight : 240.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/016754.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/016754.html
Availability details for
Quantity
  • Availability
  • Add to Cart