MAT062353543 Matrix Scientific

6-Chloro-7-methyl-1H-indole-2-carboxylic acid

  • MDL number MFCD02664432
  • Cas Number : 383132-29-4
  • Molecular Weight : 209.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/016198.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/016198.html
Availability details for
Quantity
  • Availability
  • Add to Cart