MAT062353564 Matrix Scientific

1-[2-(2-Hydroxyethylamino)-benzoimidazol-1-yl]-3,3-dimethyl-butan-2-one hydrobromide

  • MDL number MFCD00965589
  • Cas Number : 154055-56-8
  • Molecular Weight : 356.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 212-213�ec)
Country of Origin USA
Product Link http://www.matrixscientific.com/007793.html

Safety Information

MSDS http://www.matrixscientific.com/007793.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart