MAT062353568 Matrix Scientific

2-Hydroxymethyl-1H-benzoimidazole-5-carboxylicacid hydrochloride

  • MDL number MFCD00781818
  • Cas Number : 247128-20-7
  • Molecular Weight : 228.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/007929.html

Safety Information

MSDS http://www.matrixscientific.com/007929.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart