MAT062353571 Matrix Scientific

(4-Hydroxy-6-methyl-2-propylsulfanyl-pyrimidin-5-yl)-acetic acid

  • MDL number MFCD02320436
  • Cas Number : 406189-83-1
  • Molecular Weight : 242.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/016680.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/016680.html
Availability details for
Quantity
  • Availability
  • Add to Cart