MAT062353584 Matrix Scientific

3-Hydroxy-2-(quinazolin-4-ylamino)propionicacid hydrochloride

  • MDL number MFCD01909994
  • Cas Number : 1396967-25-1
  • Molecular Weight : 269.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/007833.html

Safety Information

MSDS http://www.matrixscientific.com/007833.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart