MAT062353597 Matrix Scientific

1-[2-(3-Chlorophenoxy)ethyl]piperazine

  • MDL number MFCD03999008
  • Cas Number : 401500-65-0
  • Molecular Weight : 240.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/016398.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/016398.html
Availability details for
Quantity
  • Availability
  • Add to Cart