MAT062353618 Matrix Scientific

1-(3-Iodo-pyridin-2-yl)-piperazine

  • MDL number MFCD04039872
  • Cas Number : 85386-98-7
  • Molecular Weight : 289.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 63.1-67.8
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/018221.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/018221.html
Availability details for
Quantity
  • Availability
  • Add to Cart