MAT062353640 Matrix Scientific

5-Isobutyl-2-methyl-4-pyrrolidin-1-ylmethyl-furan-3-carboxylic acid hydrochloride

  • MDL number MFCD03198371
  • Cas Number : 435341-88-1
  • Molecular Weight : 301.82

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/008635.html

Safety Information

MSDS http://www.matrixscientific.com/008635.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart