MAT062353665 Matrix Scientific

2,5-Dimethyl-thiophene-3-carbaldehyde

  • MDL number MFCD03426895
  • Cas Number : 26421-44-3
  • Molecular Weight : 140.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/016724.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/016724.html
Availability details for
Quantity
  • Availability
  • Add to Cart