MAT062353668 Matrix Scientific

4,5-Diodoveratrole

  • MDL number MFCD00094489
  • Cas Number : 110190-08-4
  • Molecular Weight : 389.96

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.matrixscientific.com/016656.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/016656.html
Availability details for
Quantity
  • Availability
  • Add to Cart