MAT062353721 Matrix Scientific

5-Isopropyl-4-methoxy-2-methyl-phenylaminehydrochloride

  • MDL number MFCD02173763
  • Cas Number : 473703-95-6
  • Molecular Weight : 215.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 250
Country of Origin USA
Product Link http://www.matrixscientific.com/010365.html

Safety Information

MSDS http://www.matrixscientific.com/010365.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart