MAT062354174 Matrix Scientific

(3-Methoxy-4-fluorobenzyl)hydrazinedihydrochloride

  • MDL number MFCD06800925
  • Cas Number : 1185081-71-3
  • Molecular Weight : 243.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/016135.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/016135.html
Availability details for
Quantity
  • Availability
  • Add to Cart