MAT062354182 Matrix Scientific

5-Methoxy-1-methyl-1H-indole-2-carbaldehyde

  • MDL number MFCD06800931
  • Cas Number : 144265-42-9
  • Molecular Weight : 189.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/016214.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/016214.html
Availability details for
Quantity
  • Availability
  • Add to Cart