MAT062354226 Matrix Scientific

(2-Methylbenzyl)hydrazine dihydrochloride

  • MDL number MFCD06800972
  • Cas Number : 1185294-85-2
  • Molecular Weight : 209.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/016140.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/016140.html
Availability details for
Quantity
  • Availability
  • Add to Cart