MAT062354240 Matrix Scientific

{[1-(2-Methylbenzyl)piperidin-4-yl]methyl}-(pyridin-3-ylmethyl)amine trihydrochloride

  • MDL number MFCD06800987
  • Cas Number : 1185303-72-3
  • Molecular Weight : 418.84

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/016433.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/016433.html
Availability details for
Quantity
  • Availability
  • Add to Cart