MAT062354245 Matrix Scientific

1'-Methyl-1,4'-bipiperidine-4-carboxylic aciddihydrochloride

  • MDL number MFCD06800993
  • Cas Number : 1185304-52-2
  • Molecular Weight : 299.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/016321.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/016321.html
Availability details for
Quantity
  • Availability
  • Add to Cart