MAT062354292 Matrix Scientific

{2-[2-(1-Methyl-1H-imidazol-2-yl)ethyl]phenyl}-amine dihydrochloride

  • MDL number MFCD06801040
  • Cas Number : 1185300-19-9
  • Molecular Weight : 274.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/016529.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/016529.html
Availability details for
Quantity
  • Availability
  • Add to Cart