MAT062354294 Matrix Scientific

[3-(2-Methyl-1H-imidazol-1-yl)propyl]aminedihydrochloride

  • MDL number MFCD06801042
  • Cas Number : 1059626-17-3
  • Molecular Weight : 212.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/016160.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/016160.html
Availability details for
Quantity
  • Availability
  • Add to Cart