MAT062354297 Matrix Scientific

[(1-Methyl-1H-indol-2-yl)methyl]amine

  • MDL number MFCD06801045
  • Cas Number : 55556-57-5
  • Molecular Weight : 160.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/016221.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/016221.html
Availability details for
Quantity
  • Availability
  • Add to Cart