MAT062354298 Matrix Scientific

N-[(1-Methyl-1H-indol-2-yl)methyl]-cyclopropanamine

  • MDL number MFCD06801046
  • Cas Number : 335031-67-9
  • Molecular Weight : 200.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/016223.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/016223.html
Availability details for
Quantity
  • Availability
  • Add to Cart