MAT062354329 Matrix Scientific

1-Methyl-2-phenylpiperazinedihydrochloride

  • MDL number MFCD06801077
  • Cas Number : 5271-28-3
  • Molecular Weight : 249.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/016468.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/016468.html
Availability details for
Quantity
  • Availability
  • Add to Cart