MAT062354365 Matrix Scientific

Morpholin-4-ylbutanoic acid hydrochloride

  • MDL number MFCD06801119
  • Cas Number : 39493-84-0
  • Molecular Weight : 209.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 131-133
Country of Origin USA
Product Link http://www.matrixscientific.com/016169.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/016169.html
Availability details for
Quantity
  • Availability
  • Add to Cart