MAT062354366 Matrix Scientific

[2-(Morpholin-4-ylmethyl)phenoxy]-acetic acid hydrochloride

  • MDL number MFCD06801121
  • Cas Number : 1185293-33-7
  • Molecular Weight : 287.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/016414.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/016414.html
Availability details for
Quantity
  • Availability
  • Add to Cart