MAT062354367 Matrix Scientific

{2-[2-(Morpholin-4-ylmethyl)phenoxy]ethyl}aminedihydrochloride

  • MDL number MFCD06801122
  • Cas Number : 1185303-97-2
  • Molecular Weight : 309.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/016417.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/016417.html
Availability details for
Quantity
  • Availability
  • Add to Cart