MAT062354368 Matrix Scientific

5-Morpholin-4-ylmethyl-2-phenyl-furan-3-carboxylic acid hydrochloride

  • MDL number MFCD06801123
  • Cas Number : 435341-95-0
  • Molecular Weight : 323.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/008632.html

Safety Information

MSDS http://www.matrixscientific.com/008632.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart